Learn More
Medchemexpress LLC Sirolimus (rapamycin) | 53123-88-9 | MFCD00867594 | 914.17 | C51H79NO13 | 50 MG
SDP
Catalog No. 5000246306
Encompass_Preferred
Rapamycin (sirolimus) GMP is a GMP-grade small-molecule mTOR inhibitor supplied as a white to off-white solid. It is intended for use as an auxiliary reagent in cell therapy manufacture and research, providing a GMP-sourced option for regulated workflows. Chemical identity is CAS 53123-88-9; formula C51H79NO13; molecular weight 914.17 g/mol.
  • Potent, specific mTOR inhibitor used in research applications.
  • GMP grade suitable for regulated cell therapy and biomanufacturing workflows.
  • White to off-white solid; handle per product documentation and SDS.
  • Chemical identity confirmed by CAS 53123-88-9.
  • Available in multiple pack sizes, including 50 mg.
Catalog No. 50-002-46306 Supplier Medchemexpress LLC Supplier No. HY10219G50MG
Rapamycin (sirolimus) GMP is a GMP-grade small-molecule mTOR inhibitor supplied as a white to off-white solid. It is intended for use as an auxiliary reagent in cell therapy manufacture and research, providing a GMP-sourced option for regulated workflows. Chemical identity is CAS 53123-88-9; formula C51H79NO13; molecular weight 914.17 g/mol.
  • Potent, specific mTOR inhibitor used in research applications.
  • GMP grade suitable for regulated cell therapy and biomanufacturing workflows.
  • White to off-white solid; handle per product documentation and SDS.
  • Chemical identity confirmed by CAS 53123-88-9.
  • Available in multiple pack sizes, including 50 mg.