Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sirolimus (rapamycin) | 53123-88-9 | MFCD00867594 | 914.17 g/mol | C51H79NO13 | 50 MG
SDP

Catalog No. 5000246943
Encompass_Preferred

Rapamycin (sirolimus) is a macrolide mTOR inhibitor supplied in a GMP-like grade for research and cell-therapy process applications. It is provided as a white to off-white solid intended for research use only and not for human therapeutic use.

  • Acts as an mTOR inhibitor supporting autophagy and signaling pathway research.
  • GMP-like grade suitable as an auxiliary reagent in cell therapy manufacture.
  • Solid, white to off-white appearance for straightforward handling and storage.
  • Molecular formula C51H79NO13 and molecular weight 914.17 g/mol for accurate formulation.
  • CAS number 53123-88-9 enables unambiguous chemical identification.

Catalog No. 50-002-46943 Supplier Medchemexpress LLC Supplier No. HY10219GL50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapamycin (sirolimus) is a macrolide mTOR inhibitor supplied in a GMP-like grade for research and cell-therapy process applications. It is provided as a white to off-white solid intended for research use only and not for human therapeutic use.

  • Acts as an mTOR inhibitor supporting autophagy and signaling pathway research.
  • GMP-like grade suitable as an auxiliary reagent in cell therapy manufacture.
  • Solid, white to off-white appearance for straightforward handling and storage.
  • Molecular formula C51H79NO13 and molecular weight 914.17 g/mol for accurate formulation.
  • CAS number 53123-88-9 enables unambiguous chemical identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.