Learn More
Medchemexpress LLC Sirolimus (rapamycin) | 53123-88-9 | MFCD00867594 | 99.9% | 914.17 | C51H79NO13 | 10 MG
SDP
Catalog No. 5000246307
Encompass_Preferred
Rapamycin (sirolimus) is a potent and specific mTOR inhibitor used as a research reagent to modulate autophagy and immune signaling. It binds FKBP12 and allosterically inhibits mTORC1, and is supplied as a high-purity small molecule for biochemical and cell-based studies.
  • Potent mTOR inhibitor; binds FKBP12 and inhibits mTORC1.
  • Activates autophagy and exhibits immunosuppressant activity.
  • High purity (99.94%) suitable for research and analytical use.
  • Soluble in DMSO (125 mg/mL) and ethanol (50 mg/mL); insoluble in water.
  • Stable as a powder at -20°C (up to 3 years); in solution store at -80°C for long-term stability.
  • Available in GMP and GMP-like variants and multiple packaging sizes.
Catalog No. 50-002-46307 Supplier Medchemexpress LLC Supplier No. HY10219GL10MG
Rapamycin (sirolimus) is a potent and specific mTOR inhibitor used as a research reagent to modulate autophagy and immune signaling. It binds FKBP12 and allosterically inhibits mTORC1, and is supplied as a high-purity small molecule for biochemical and cell-based studies.
  • Potent mTOR inhibitor; binds FKBP12 and inhibits mTORC1.
  • Activates autophagy and exhibits immunosuppressant activity.
  • High purity (99.94%) suitable for research and analytical use.
  • Soluble in DMSO (125 mg/mL) and ethanol (50 mg/mL); insoluble in water.
  • Stable as a powder at -20°C (up to 3 years); in solution store at -80°C for long-term stability.
  • Available in GMP and GMP-like variants and multiple packaging sizes.