Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sirolimus (rapamycin) | 53123-88-9 | MFCD00867594 | 99.9% | 914.17 g/mol | C51H79NO13 | 5 MG
SDP

Catalog No. 5000245828
Encompass_Preferred

Rapamycin (sirolimus) is a macrocyclic lactone that potently and specifically inhibits mTOR; it is used in research to modulate autophagy and mTOR-dependent signaling. Supplied as a research-grade powder, it is suitable for preparing stock solutions and for use in in vitro and in vivo formulations.

  • Powder form for laboratory use.
  • Molecular formula C51H79NO13.
  • Molecular weight 914.17 g/mol.
  • CAS number 53123-88-9.
  • Soluble in DMSO at 100 mg/mL (needs ultrasonic).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Potent mTOR inhibitor (IC50 0.1 nM in HEK293 cells).
  • Available in small pack sizes suitable for stock preparation, including 5 mg.

Catalog No. 50-002-45828 Supplier Medchemexpress LLC Supplier No. HY102195MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapamycin (sirolimus) is a macrocyclic lactone that potently and specifically inhibits mTOR; it is used in research to modulate autophagy and mTOR-dependent signaling. Supplied as a research-grade powder, it is suitable for preparing stock solutions and for use in in vitro and in vivo formulations.

  • Powder form for laboratory use.
  • Molecular formula C51H79NO13.
  • Molecular weight 914.17 g/mol.
  • CAS number 53123-88-9.
  • Soluble in DMSO at 100 mg/mL (needs ultrasonic).
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Potent mTOR inhibitor (IC50 0.1 nM in HEK293 cells).
  • Available in small pack sizes suitable for stock preparation, including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.