Learn More
Sino Biological SIRP alpha / CD172a Antibody (APC), Rabbit MAb 100Tests
Supplier:  Sino Biological 50956R001A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIRP alpha / CD172a (rM SIRP alpha / CD172a; Catalog#50956-M08H; BAA20376.1; Met1-Asn373) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-222-3301