Learn More
Sino Biological SIRP alpha / CD172a Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11612RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SIRPA (rh SIRPA; Catalog#11612-H08H; NP_542970.1; Met1-Arg370, Gly 75 Ala). SIRPA specific IgG was purified by human SIRPA affinity chromatography.
Catalog No. 50-222-3272