Learn More
Sino Biological Human SIRP alpha Protein (ECD, Fc Tag) 50ug
Supplier:  Sino Biological 11612H02H150
Encompass_Preferred
A DNA sequence encoding the human SIRPA (NP_542970.1) (Met1-Arg370) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5991