Learn More
Sino Biological Human SIRP alphaV2 Protein (ECD, His Tag) 100ug
Supplier:  Sino Biological 30014H08H100
Encompass_Preferred
A DNA sequence encoding the human SIRP alphaV2 (AAH38510.1) (Met1-Arg369) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6006