Learn More
Medchemexpress LLC SIRP beta 1 Human 100ug
SDP
Catalog No. 5000185089
Encompass_Preferred
Recombinant human SIRP beta 1 protein expressed in HEK293 cells with a C-terminal 6xHis tag, supplied lyophilized from PBS (pH 7.4). The product is provided at 45-55 kDa and >95% purity for use in biochemical, signaling, and protein-protein interaction studies involving SIRPB1 and SYK recruitment. Reconstitute according to guidance to ensure stability.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • HEK293-expressed human protein for native-like post-translational modifications.
  • C-terminal 6xHis tag for straightforward purification and detection.
  • Lyophilized from PBS (pH 7.4) for long-term storage stability.
  • Suitable for signaling, interaction, and biochemical assays.
  • Reconstitute to ≥100 μg/mL in ddH2O to maintain protein stability.
Catalog No. 50-001-85089 Supplier Medchemexpress LLC Supplier No. HYP70719100UG
Recombinant human SIRP beta 1 protein expressed in HEK293 cells with a C-terminal 6xHis tag, supplied lyophilized from PBS (pH 7.4). The product is provided at 45-55 kDa and >95% purity for use in biochemical, signaling, and protein-protein interaction studies involving SIRPB1 and SYK recruitment. Reconstitute according to guidance to ensure stability.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • HEK293-expressed human protein for native-like post-translational modifications.
  • C-terminal 6xHis tag for straightforward purification and detection.
  • Lyophilized from PBS (pH 7.4) for long-term storage stability.
  • Suitable for signaling, interaction, and biochemical assays.
  • Reconstitute to ≥100 μg/mL in ddH2O to maintain protein stability.