Learn More
ACROBiosystems ACROBiosystems Biotinylated Human SIRP alpha / CD172a Protein, His,Avitag™ (HPLC-verified)
Catalog No. 501957119
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (SIA-H82E0) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession # NP_001035111).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 39.9 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (Cat. No. CD7-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (Cat. No. SIA-H82E0) with a linear range of 0.078-0.625 μg/mL (QC tested).
Catalog No. 50-195-7119 Supplier ACROBiosystems Supplier No. SIAH82E025UG
Recombinant Protein;25UG;Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (SIA-H82E0) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 370 (Accession # NP_001035111).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 39.9 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (Cat. No. CD7-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alpha, His,Avitag (HPLC-verified) (Cat. No. SIA-H82E0) with a linear range of 0.078-0.625 μg/mL (QC tested).