Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems SIRP Alpha Protein, Fc Tag, Cynomolgus, Lyophilized, 1 mg

Catalog No. 501657711
Encompass_Preferred

This Cynomolgus SIRP alpha (CD172a) protein is a recombinant, Fc-tagged immunoglobulin-like cell surface receptor expressed in human 293 cells. As a member of the SIRP family, it acts as a docking protein and negative regulator of signaling pathways involved in phagocytosis and immune cell activation. This product is supplied as a lyophilized powder verified by SEC-MALS.

  • Greater than 95% purity by SDS-PAGE and greater than 90% purity by SEC-MALS
  • Contains human IgG1 Fc tag at the C-terminus
  • Calculated molecular weight of 63.6 kDa, migrating as 70 to 85 kDa under reducing conditions
  • Formulated in Tris with glycine, arginine, and NaCl, pH 7.5 with trehalose as protectant
  • Endotoxin levels less than 1.0 EU per μg
  • Compatible with binding assays for CD47

Catalog No. 50-165-7711 Supplier ACROBiosystems Supplier No. SIAC52541MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Cynomolgus SIRP alpha (CD172a) protein is a recombinant, Fc-tagged immunoglobulin-like cell surface receptor expressed in human 293 cells. As a member of the SIRP family, it acts as a docking protein and negative regulator of signaling pathways involved in phagocytosis and immune cell activation. This product is supplied as a lyophilized powder verified by SEC-MALS.

  • Greater than 95% purity by SDS-PAGE and greater than 90% purity by SEC-MALS
  • Contains human IgG1 Fc tag at the C-terminus
  • Calculated molecular weight of 63.6 kDa, migrating as 70 to 85 kDa under reducing conditions
  • Formulated in Tris with glycine, arginine, and NaCl, pH 7.5 with trehalose as protectant
  • Endotoxin levels less than 1.0 EU per μg
  • Compatible with binding assays for CD47

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.