Learn More
ACROBiosystems Recombinant Protein;Mouse SIRP alpha / CD172a Protein, Fc Tag;HEK293;1MG;SIA,SIRP alpha,SHPS1,SIRPA,CD172A,BIT,SHPS-1,MFR,MYD-1,MYD1,PTPNS1,SIRPalpha,p84,SIRP-alpha,CD172
Supplier:  ACROBiosystems SIAM52581MG
Encompass_Preferred
Recombinant Protein;Mouse SIRP alpha / CD172a Protein, Fc Tag;1MG;Mouse SIRP alpha, Fc Tag (SIA-M5258) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # P97797.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.3 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-M525a) at 10 μg/mL (100 μL/well) can bind Mouse SIRP alpha, Fc Tag (Cat. No. SIA-M5258) with a linear range of 10-156 ng/ml (QC tested).
Catalog No. 50-165-7652