Learn More
ACROBiosystems Biotinylated Human SIRP Alpha Protein, Fc, Avitag™, 25 μg
Catalog No. 501658394
Encompass_Preferred
This biotinylated human Signal-regulatory protein alpha (SIRP alpha), also known as CD172a, is a member of the immunoglobulin superfamily expressed from human HEK293 cells. It serves as an immunoglobulin-like cell surface receptor for CD47 and acts as a docking protein that induces the translocation of binding partners to the plasma membrane, playing a role in signaling, phagocytosis, and dendritic cell activation. This protein is C-terminally tagged with a human IgG1 Fc tag followed by an Avi tag.
  • Greater than 95% purity as determined by SDS-PAGE
  • Greater than 90% purity as determined by SEC-MALS
  • Calculated molecular weight of 65.8 kDa, migrating at 75 to 105 kDa under reducing conditions due to glycosylation
  • Supplied as a 0.2 μm filtered solution in Tris with glycine, arginine, and NaCl at pH 7.5 with trehalose
  • Enzymatically biotinylated using Avi tag technology
Catalog No. 50-165-8394 Supplier ACROBiosystems Supplier No. CDAH82F225UG
This biotinylated human Signal-regulatory protein alpha (SIRP alpha), also known as CD172a, is a member of the immunoglobulin superfamily expressed from human HEK293 cells. It serves as an immunoglobulin-like cell surface receptor for CD47 and acts as a docking protein that induces the translocation of binding partners to the plasma membrane, playing a role in signaling, phagocytosis, and dendritic cell activation. This protein is C-terminally tagged with a human IgG1 Fc tag followed by an Avi tag.
  • Greater than 95% purity as determined by SDS-PAGE
  • Greater than 90% purity as determined by SEC-MALS
  • Calculated molecular weight of 65.8 kDa, migrating at 75 to 105 kDa under reducing conditions due to glycosylation
  • Supplied as a 0.2 μm filtered solution in Tris with glycine, arginine, and NaCl at pH 7.5 with trehalose
  • Enzymatically biotinylated using Avi tag technology