Learn More
ACROBiosystems Recombinant Protein;Cynomolgus SIRP alpha / CD172a Protein;HEK293;100UG;SIA,SIRP alpha,SHPS1,SIRPA,CD172A,BIT,SHPS-1,MFR,MYD-1,MYD1,PTPNS1,SIRPalpha,p84,SIRP-alpha,CD172
Supplier:  ACROBiosystems SIAC52H7100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus SIRP alpha / CD172a Protein;100UG;Cynomolgus SIRP alpha (SIA-C52H7) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 370 (Accession # NP_001271679.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.0 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque CD47, Fc Tag (Cat. No. CD7-C5252) at 2 μg/mL (100 μL/well) can bind Cynomolgus SIRP alpha (Cat. No. SIA-C52H7) with a linear range of 0.313-2.5 μg/mL (QC tested).
Catalog No. 50-165-7783