Learn More
ACROBiosystems Recombinant Protein;Human SIRP beta / CD172b Protein, Fc Tag;HEK293;1MG;SIA,9930027N05Rik,SIRP-beta,Sirpb,Sirpb1,CD172 antigen-like family member B,CD172b antigen,CD172b,DKFZp686A05192,FLJ26614,SIRP beta 1,SIRPB1,SIRP-BETA-1,SIRP beta
Catalog No. 501657709
Encompass_Preferred
Recombinant Protein;Human SIRP beta / CD172b Protein, Fc Tag;1MG;Human SIRP beta, Fc Tag (SIA-H5257) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Leu 371 (Accession # O00241-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.7 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7709 Supplier ACROBiosystems Supplier No. SIAH52571MG
Recombinant Protein;Human SIRP beta / CD172b Protein, Fc Tag;1MG;Human SIRP beta, Fc Tag (SIA-H5257) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Leu 371 (Accession # O00241-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.7 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.