Learn More
Medchemexpress LLC HY-100591 5mg Medchemexpress, SirReal2 CAS:709002-46-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1005915MG
Encompass_Preferred
Medchemexpress, HY-100591 5mg SirReal2 CAS:709002-46-0 SirReal2 is a potent, isotype-selective Sirt2 inhibitor with an IC50 value of 140 nM and has very little effect on the activities of Sirt3-5. SirReal2 leads to tubulin hyperacetylation in HeLa cells and induces destabilization of the checkpoint protein BubR1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7754