Learn More
Enzo Life Sciences FLUOR DE LYS SIRT1 fluorometric drug discovery assay kit (1x96wells)
SDP
Catalog No. NC9209259
Encompass_Preferred
First-to-market by the leader in Epigenetics research tools. A FLUOR DE LYS. SIRT1 assay is based on the FLUOR DE LYS SIRT1 Substrate and FLUOR DE LYS Developer II combination. The assay procedure has two steps. First, the FLUOR DE LYS SIRT1 Substrate, which contains a peptide comprising amino acids 379-382 of human p53 (Arg-His-Lys-Lys(Ac)). Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the FLUOR DE LYSDeveloper II produces a fluorophore. Alternative Name: Sirtuin 1 fluorescent assay kit. Applications: Fluorescent detection, HTS, Cell-based assays, Activity assay.
Catalog No. NC9209259 Supplier Enzo Life Sciences Supplier No. BMLAK5550001
First-to-market by the leader in Epigenetics research tools. A FLUOR DE LYS. SIRT1 assay is based on the FLUOR DE LYS SIRT1 Substrate and FLUOR DE LYS Developer II combination. The assay procedure has two steps. First, the FLUOR DE LYS SIRT1 Substrate, which contains a peptide comprising amino acids 379-382 of human p53 (Arg-His-Lys-Lys(Ac)). Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the FLUOR DE LYSDeveloper II produces a fluorophore. Alternative Name: Sirtuin 1 fluorescent assay kit. Applications: Fluorescent detection, HTS, Cell-based assays, Activity assay.