Learn More
Enzo Life Sciences SIRT1 (human) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4270100
Encompass_Preferred
Immunogen: Recombinant human SIRT1 (sirtuin 1). Source: Purified from rabbit serum. Species reactivity: Human. Specificity: Recognizes recombinant and native SIRT1. Applications: IHC (PS), WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS, pH 7.4.
Catalog No. 50-201-1821