Learn More
Enzo Life Sciences FLUOR DE LYS SIRT1 deacetylase substrate (0.5 µmol)
SDP
Catalog No. NC9947405
Encompass_Preferred
Formulation: Supplied as a 5 mM solution in HDAC Assay Buffer. Purity: ≥95% (HPLC). Long Term Storage: -80°C
Catalog No. NC9947405 Supplier Enzo Life Sciences Supplier No. BMLKI1770005
Formulation: Supplied as a 5 mM solution in HDAC Assay Buffer. Purity: ≥95% (HPLC). Long Term Storage: -80°C