Learn More
Cayman Chemical SIRT1/2 Inhibitor IV, Purity: ≥98%, CAS: 14513-15-6, 5 mg
SDP
Catalog No. 502969157
Encompass_Preferred
A cell-permeable inhibitor of SIRT1 (IC50 = 56 µM) and SIRT2 (IC50 = 59 µM); less effectively inhibits SIRT5 (IC50 >300 µM) and has no effect on class I and II HDACs; sensitizes H460 lung cancer cells to etoposide and paclitaxel; blocks a SIRT1-dependent hypoxic response in vivo
Catalog No. 50-296-9157 Supplier Cayman Chemical Supplier No. 144075 MG
A cell-permeable inhibitor of SIRT1 (IC50 = 56 µM) and SIRT2 (IC50 = 59 µM); less effectively inhibits SIRT5 (IC50 >300 µM) and has no effect on class I and II HDACs; sensitizes H460 lung cancer cells to etoposide and paclitaxel; blocks a SIRT1-dependent hypoxic response in vivo