Learn More
Medchemexpress LLC Sirt2/6-in-1 (compound 5) | 1214468-35-5 | MFCD11667663 | 99.8% | 410.40 g·mol⁻¹ | C19H14N4O5S | 5MG
SDP
Catalog No. 5000238079
Encompass_Preferred
SIRT2/6-IN-1 is a small-molecule research inhibitor that targets sirtuin deacetylases SIRT6 and SIRT2. It inhibits both enzymes with micromolar potency and has demonstrated cellular effects including increased H3K9 acetylation, enhanced glucose uptake, and reduced TNF-α secretion. The compound is supplied as a powder for laboratory research and is soluble in DMSO and several in vivo vehicle formulations.
Catalog No. 50-002-38079 Supplier Medchemexpress LLC Supplier No. HY1231945MG
SIRT2/6-IN-1 is a small-molecule research inhibitor that targets sirtuin deacetylases SIRT6 and SIRT2. It inhibits both enzymes with micromolar potency and has demonstrated cellular effects including increased H3K9 acetylation, enhanced glucose uptake, and reduced TNF-α secretion. The compound is supplied as a powder for laboratory research and is soluble in DMSO and several in vivo vehicle formulations.