Learn More
Aviva Systems Biology Corp SIRT2 Antibody - C-terminal region (ARP32385_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32385_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SIRT2. Alternate names: SIR2, SIR2L, SIR2L2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54559