Learn More
Enzo Life Sciences SIRT2 (human), (recombinant) (His-tag) (500 U)
SDP
Supplier:  Enzo Life Sciences BMLSE2510500
Encompass_Preferred
Alternative name: Sirtuin 2. Formulation: Liquid. In 25mM TRIS, pH 7.5, containing 100mM NaCl, 5mM DTT and 10% glycerol. Source: Produced in E. coli. Contains a N-terminal His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-1961