Learn More
Medchemexpress LLC HY-112427 10mg Medchemexpress, Sirt2-IN-1 CAS:1862238-00-3 Purity:>98%
SDP
Catalog No. 501969088
Encompass_Preferred
Medchemexpress, HY-112427 10mg Sirt2-IN-1 CAS:1862238-00-3 Sirt2-IN-1 (Compound 9) is a sirtuin 2 (Sirt2) inhibitor with an IC50 of 163 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9088 Supplier Medchemexpress LLC Supplier No. HY11242710MG
Medchemexpress, HY-112427 10mg Sirt2-IN-1 CAS:1862238-00-3 Sirt2-IN-1 (Compound 9) is a sirtuin 2 (Sirt2) inhibitor with an IC50 of 163 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.