Learn More
Medchemexpress LLC Sirt2-in-8 | 115687-05-3 | 99.3% | 396.83 g·mol⁻1 | C20H17ClN4O3 | 10 MG
SDP
Catalog No. 5000241463
Encompass_Preferred
SIRT2-IN-8 is a selective, cell-permeable small-molecule inhibitor of the SIRT2 deacetylase enzyme used in preclinical research to probe sirtuin-regulated pathways and neurodegenerative disease models, including Huntington's and Parkinson's disease studies.
  • Selective SIRT2 inhibition for sirtuin pathway studies.
  • Reported purity 99.32%.
  • Molecular formula C20H17ClN4O3 and molecular weight 396.83 g·mol⁻¹.
  • Available in research-scale quantities (5-100 mg).
  • Datasheet includes solubility and handling recommendations for laboratory use.
Catalog No. 50-002-41463 Supplier Medchemexpress LLC Supplier No. HY10766010MG
SIRT2-IN-8 is a selective, cell-permeable small-molecule inhibitor of the SIRT2 deacetylase enzyme used in preclinical research to probe sirtuin-regulated pathways and neurodegenerative disease models, including Huntington's and Parkinson's disease studies.
  • Selective SIRT2 inhibition for sirtuin pathway studies.
  • Reported purity 99.32%.
  • Molecular formula C20H17ClN4O3 and molecular weight 396.83 g·mol⁻¹.
  • Available in research-scale quantities (5-100 mg).
  • Datasheet includes solubility and handling recommendations for laboratory use.