Learn More
Aviva Systems Biology Corp SIRT3 Antibody - C-terminal region (ARP32389_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32389_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SIRT3. Alternate names: SIR2L3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54564