Learn More
Aviva Systems Biology Corp SIRT3 Antibody - C-terminal region (ARP32389_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32389_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SIRT3. Alternate names: SIR2L3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54563