Learn More
Cayman Chemical SIRT3 Direct Fluorescent Scre
SDP
Catalog No. 502886850
Encompass_Preferred
Cayman’s SIRT3 Direct Fluorescent Screening Assay Kit provides a convenient fluorometric method for screening SIRT3 modulators in a two-step reaction. In the first step, the SIRT2/SIRT3 peptide and NAD+ are incubated with human recombinant SIRT3. Deacetylation sensitizes the substrate such that treatment with the developer in the second step releases a fluorescent product. The fluorophore can be EAsily analyzed using an excitation wavelength of 360 nm and an emission wavelength of 465 nm.
Catalog No. 50-288-6850 Supplier Cayman Chemical Supplier No. 1001156696 WELL
Cayman’s SIRT3 Direct Fluorescent Screening Assay Kit provides a convenient fluorometric method for screening SIRT3 modulators in a two-step reaction. In the first step, the SIRT2/SIRT3 peptide and NAD+ are incubated with human recombinant SIRT3. Deacetylation sensitizes the substrate such that treatment with the developer in the second step releases a fluorescent product. The fluorophore can be EAsily analyzed using an excitation wavelength of 360 nm and an emission wavelength of 465 nm.