Learn More
Sino Biological Recombinant Mouse KDM5B Protein (GST Tag)
Catalog No. 502574500
Encompass_Preferred
Recombinant mouse KDM5B (1-753) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4500 Supplier Sino Biological Supplier No. K42531BG1000
Recombinant mouse KDM5B (1-753) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.