Learn More
Aviva Systems Biology Corp SIRT4 Antibody - middle region (ARP32451_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32451_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SIRT4. Alternate names: MGC130046, MGC130047, MGC57437, SIR2L4, sirtuin 4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54598