Learn More
Sino Biological Recombinant Human KDM5C Protein (GST Tag)
Catalog No. 502574498
Encompass_Preferred
Recombinant human KDM5C (1-671) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4498 Supplier Sino Biological Supplier No. K42531CG1000
Recombinant human KDM5C (1-671) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.