Learn More
Aviva Systems Biology Corp SIRT5 Antibody - C-terminal region (ARP40125_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40125_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SIRT5. Alternate names: SIR2L5ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57301