Learn More
Aviva Systems Biology Corp SIRT5 Antibody - middle region (ARP32391_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32391_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human SIRT5. Alternate names: SIR2L5ApplicationIHC, WB. Species reactivity: Cow, Goat, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-54566