Learn More
Aviva Systems Biology Corp SIRT5 Antibody - middle region (ARP40126_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40126_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SIRT5. Alternate names: SIR2L5ApplicationWB, IHC. Species reactivity: Cow, Goat, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-57302