Learn More
Aviva Systems Biology Corp SIRT5 Antibody - C-terminal region (ARP32390_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32390_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SIRT5. Alternate names: SIR2L5ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54565