Learn More
Enzo Life Sciences SIRT5 (human), (recombinant) (His-tag) (50 kU)
SDP
Supplier:  Enzo Life Sciences BMLSE5550050
Encompass_Preferred
Alternative name: Sirtuin 5, Sir2 like protein 5, SIR2L5, NAD dependent deacetylase Sirtuin 5. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 25mM TRIS, pH 7.5, 100mM sodium chloride, 5 mM DTT, containing 10% (v/v) glycerol. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2088