Learn More
Medchemexpress LLC HY-112634 10mg Medchemexpress, SIRT5 inhibitor 1 CAS:2166487-21-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11263410MG
Encompass_Preferred
Medchemexpress, HY-112634 10mg SIRT5 inhibitor 1 CAS:2166487-21-2 SIRT5 inhibitor 1 is a potent Human Sirtuin 5 deacylase inhibitor, with an IC50 of 0.11 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9125