Learn More
Sino Biological Recombinant Human PPME1 Protein (GST Tag)
Catalog No. 502573720
Encompass_Preferred
Recombinant human mature form of PPME1 (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3720 Supplier Sino Biological Supplier No. P33230G1000
Recombinant human mature form of PPME1 (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.