Learn More
Medchemexpress LLC SIRT6 activator 12q | 2601734-99-8 | 99.8% | 454.52 | 5 MG
SDP
Catalog No. 5000362305
Encompass_Preferred
SIRT6 activator 12q is a potent, selective, and orally active SIRT6 activator with an IC50 of 0.58 μM for SIRT6. It inhibits cell growth and migration, induces apoptosis and cell cycle arrest at the G2 phase, and demonstrates anticancer activity.
  • Inhibits cell growth and migration
  • Induces apoptosis and cell cycle arrest at the G2 phase
  • Demonstrates anticancer activity
  • Shows antiproliferative activity against human pancreatic cancer cell lines (PANC-1, BXPC-3, MIA PaCa-2, and AsPC-1)
  • Decreases protein expression of H3K9ac, H3K18ac, and H3K56ac in PANC-1 and BXPC-3 cells
Catalog No. 50-003-62305 Supplier Medchemexpress LLC Supplier No. HY1487125MG
SIRT6 activator 12q is a potent, selective, and orally active SIRT6 activator with an IC50 of 0.58 μM for SIRT6. It inhibits cell growth and migration, induces apoptosis and cell cycle arrest at the G2 phase, and demonstrates anticancer activity.
  • Inhibits cell growth and migration
  • Induces apoptosis and cell cycle arrest at the G2 phase
  • Demonstrates anticancer activity
  • Shows antiproliferative activity against human pancreatic cancer cell lines (PANC-1, BXPC-3, MIA PaCa-2, and AsPC-1)
  • Decreases protein expression of H3K9ac, H3K18ac, and H3K56ac in PANC-1 and BXPC-3 cells