Learn More
Sino Biological Recombinant Human SIRT1 Protein (GST Tag), Active
Catalog No. 502573061
Encompass_Preferred
Recombinant human SIRT1 (193-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3061 Supplier Sino Biological Supplier No. S3531G50
Recombinant human SIRT1 (193-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.