Learn More
Aviva Systems Biology Corp SIRT6 Antibody - N-terminal region (ARP32409_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32409_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SIRT6. Alternate names: SIR2L6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54578