Learn More
TARGETMOL CHEMICALS INC SIRT6-IN-5 5MG
SDP
Catalog No. 502580782
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SIRT6-IN-5(SIRT6 inhibitor 5) is a potent and selective SIRT6 inhibitor with an IC50 value of 34 uM.SIRT6-IN-5 is immunosuppressive and chemo-sensitizing increases H3K9 acetylation and glucose uptake in cultured cells and reduces T-cell proliferation. purity: 98%
Catalog No. 50-258-0782 Supplier TARGETMOL CHEMICALS INC Supplier No. T247935MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. SIRT6-IN-5(SIRT6 inhibitor 5) is a potent and selective SIRT6 inhibitor with an IC50 value of 34 uM.SIRT6-IN-5 is immunosuppressive and chemo-sensitizing increases H3K9 acetylation and glucose uptake in cultured cells and reduces T-cell proliferation. purity: 98%