Learn More
Aviva Systems Biology Corp SIRT7 Antibody - middle region (ARP32405_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32405_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIRT7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SIRT7. Alternate names: MGC126840, MGC126842, SIR2L7ApplicationWB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-014-54575