Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sirt7 inhibitor 97491 | 1807758-81-1 | 98.8% | C15H12ClN3O | 10MG
SDP

Catalog No. 5000260515
Encompass_Preferred

Sirt7 inhibitor 97491 is a small-molecule research reagent that selectively inhibits SIRT7 deacetylase activity, increases p53 acetylation at K373/382, and promotes apoptosis via caspase pathways. It is intended for biochemical and cell-based studies of SIRT7 function and related signaling pathways.

  • Potent SIRT7 inhibition (IC50 325 nM).
  • High purity (98.8%), suitable for research applications.
  • Molecular formula C15H12ClN3O; molecular weight 285.73 g·mol⁻¹.
  • Supplied as a solid, off-white to light yellow appearance.
  • Storage: 4°C protected from light; in solution, -80°C (6 months) or -20°C (1 month).
  • Available as mg-scale powders and DMSO solution for assays.

Catalog No. 50-002-60515 Supplier Medchemexpress LLC Supplier No. HY13589910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sirt7 inhibitor 97491 is a small-molecule research reagent that selectively inhibits SIRT7 deacetylase activity, increases p53 acetylation at K373/382, and promotes apoptosis via caspase pathways. It is intended for biochemical and cell-based studies of SIRT7 function and related signaling pathways.

  • Potent SIRT7 inhibition (IC50 325 nM).
  • High purity (98.8%), suitable for research applications.
  • Molecular formula C15H12ClN3O; molecular weight 285.73 g·mol⁻¹.
  • Supplied as a solid, off-white to light yellow appearance.
  • Storage: 4°C protected from light; in solution, -80°C (6 months) or -20°C (1 month).
  • Available as mg-scale powders and DMSO solution for assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.