Learn More
ABclonal Technology SIRT7 Rabbit mAb
SDP
Catalog No. 5000007360
Encompass_Preferred
This gene encodes a member of the sirtuin family of proteins homologs to the yeast Sir2 protein Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes The functions of human sirtuins have not yet been determined however yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity The protein encoded by this gene is included in class IV of the sirtuin family
Catalog No. 50-000-07360 Supplier ABclonal Technology Supplier No. A22735100UL
This gene encodes a member of the sirtuin family of proteins homologs to the yeast Sir2 protein Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes The functions of human sirtuins have not yet been determined however yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity The protein encoded by this gene is included in class IV of the sirtuin family