Learn More
Enzo Life Sciences Sirtinol (1mg). CAS: 410536-97-9
SDP
Supplier:  Enzo Life Sciences ALX270308M001
Encompass_Preferred
SIRT inhibitor. Specific cell permeable inhibitor of the sirtuin family of NAD-dependent deacetylases (ySir2: IC50=48µM; hSIRT1: IC50=60µM; hSIRT2: IC50=58µM) with no effect on human HDAC1. Reported to inhibit Sir2p transcriptional silencing activity in vivo (IC50=25µM) and NAD-dependent histone deacetylase activity of purified recombinant yeast Sir2p (IC50=70µM) and hSIRT2 (IC50=40µM) in vitro. Alternative name: Sir two inhibitor naphthol, 2-[(2-Hydroxynaphthalen-1-yl-methylene)amino]-N-(1-phenethyl)benzamide. Formula: C26H22N2O2. MW: 394.5. Purity: ≥97% (HPLC). Appearance: Off-white to yellow crystals. Solubility: Soluble in DMSO, 100% ethanol or dimethyl formamide. Use/Stability: Relatively unstable in aqueous solution, especially at acidic pH. Long Term Storage: +4°C. Handling: Protect from moisture. Protect from light. Store under inert gas.
Catalog No. 50-200-8480