Learn More
TARGETMOL CHEMICALS INC SIRTINOL 25MG
SDP
Catalog No. 502579311
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sirtinol is a selective SIRT1/2 inhibitor (IC50 131/38 uM). purity: 99%
Catalog No. 50-257-9311 Supplier TARGETMOL CHEMICALS INC Supplier No. T667125MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sirtinol is a selective SIRT1/2 inhibitor (IC50 131/38 uM). purity: 99%