Learn More
AdipoGen Sirtinol (25 mg)
SDP
Supplier:  AdipoGen AGCR10055M025
Encompass_Preferred
Chemical. CAS: 410536-97-9. Formula: C26H22N2O2. MW: 394.5. Specific cell permeable SIRT1 (sirtuin 1) inhibitor. Senescence-like growth arrest inducer. Platelet aggregation inhibitor. Apoptosis inducer.
Catalog No. 50-205-3811