Learn More
Eurofins Discoverx Sirtuin 1, 50 µg. (EPI059)
Supplier:  Eurofins Discoverx EPI059
Encompass
Sirtuin 1 is purified, N terminal 6xHis tagged recombinant Sirtuin 1 enzyme. This enzyme contains amino acids 50-356. It retains its binding/enzymatic activity and is intended for use in epigenetic assays. This enzyme is provided as 1 mg/mL frozen in 50% glycerol. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-200-5291