Learn More
AdipoGen Sirtuin 2 human pAb
SDP
Supplier:  AdipoGen AG25A0083C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human sirtuin 2. Detects a band of ~45kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Sirtuin-2 is a NAD-dependent protein deacetylase, which deacetylates the Lys-40 of a-tubulin. It is involved in the control of mitotic exit in the cell cycle. Furthermore, a variety of age-associated diseases such as type 2 diabetes, obesity, osteoporosis, and Alzheimer’s disease are associated with sirtuins. Selective inhibition of Sirtuin-2 achieves neuroprotection and identifies Sirtuin-2 to be a potential therapeutic avenue in Huntington’s and Parkinson’s disease.
Catalog No. 50-256-3234